Learn More
Cayman Chemical PAF C-16-d4 100ug
SDP
Catalog No. 502911828
Encompass_Preferred
An internal standard for the quantification of PAF C-16 by GC- or LC-MS.
Catalog No. 50-291-1828 Supplier Cayman Chemical Supplier No. 360900100 UG
An internal standard for the quantification of PAF C-16 by GC- or LC-MS.