Learn More
Cayman Chemical PAF C-16-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 211106-54-6, 1 mg
SDP
Catalog No. 502911980
Encompass_Preferred
An internal standard for the quantification of PAF C-16 by GC- or LC-MS.
Catalog No. 50-291-1980 Supplier Cayman Chemical Supplier No. 3609001 MG
An internal standard for the quantification of PAF C-16 by GC- or LC-MS.