Learn More
Cayman Chemical PAF C-16-d4 500ug
SDP
Catalog No. 502909248
Encompass_Preferred
An internal standard for the quantification of PAF C-16 by GC- or LC-MS.
Catalog No. 50-290-9248 Supplier Cayman Chemical Supplier No. 360900500 UG
An internal standard for the quantification of PAF C-16 by GC- or LC-MS.