Learn More
Cayman Chemical PAF C-18-d4, Purity: ≥99% deuterated forms (d1-d4), 1 mg
SDP
Catalog No. 502891056
Encompass_Preferred
An internal standard for the quantification of PAF C-18 by GC- or LC-MS
Catalog No. 50-289-1056 Supplier Cayman Chemical Supplier No. 100102291 MG
An internal standard for the quantification of PAF C-18 by GC- or LC-MS