Learn More
Cayman Chemical PAF Receptr Blockng PeptID 1ea
SDP
Catalog No. 502911632
Encompass_Preferred
An inhibitor of microtubule assembly cytotoxic to a variety of cancer cells including SKOV3 ovarian U87 glioblastoma and AsPC-1 pancreatic cancer cells (IC50s 36 77 and 285 nM respectively) induces complete tumor regression in a rat Yoshida AH13 sarcoma model at 10 mg/kg without inducing systemic or neurotoxicity
Catalog No. 50-291-1632 Supplier Cayman Chemical Supplier No. 3606001 EA
An inhibitor of microtubule assembly cytotoxic to a variety of cancer cells including SKOV3 ovarian U87 glioblastoma and AsPC-1 pancreatic cancer cells (IC50s 36 77 and 285 nM respectively) induces complete tumor regression in a rat Yoshida AH13 sarcoma model at 10 mg/kg without inducing systemic or neurotoxicity