Learn More
Cayman Chemical PAF Receptor (human) Monoclonal Antibody (11A4, Clone 21), 1 ea
SDP
Catalog No. 502973212
Encompass_Preferred
Immunogen: Synthetic peptide from the internal region of human PAF receptor • Host: Mouse, 11A4 clone 21 • Species Reactivity: (+) Human, bovine, and porcine PAF receptor • Isotype: IgG2a • Application(s): ELISA, FC, and ICC; does not work for WB • PAF is a biologically active phospholipid whose biological effects include activation of platelets, polymorphonuclear leukocytes, monocytes, and macrophages. PAF increases vascular permeability, decreases cardiac output, induces hypotension, and stimulates uterine contraction.
Catalog No. 50-297-3212 Supplier Cayman Chemical Supplier No. 1606001 EA
Immunogen: Synthetic peptide from the internal region of human PAF receptor • Host: Mouse, 11A4 clone 21 • Species Reactivity: (+) Human, bovine, and porcine PAF receptor • Isotype: IgG2a • Application(s): ELISA, FC, and ICC; does not work for WB • PAF is a biologically active phospholipid whose biological effects include activation of platelets, polymorphonuclear leukocytes, monocytes, and macrophages. PAF increases vascular permeability, decreases cardiac output, induces hypotension, and stimulates uterine contraction.