Learn More
Cayman Chemical PAF RECPTR Blockng PeptID
SDP
Catalog No. 502972328
Encompass_Preferred
A natural 14-naphthoquinone that has diverse effects in cells and animals causes the generation of ROS and induces apoptosis in cancer cells activates signaling through Nrf2 and ARE inducing the expression of Nrf2 target genes including NQO1 and HO-1 in cultured neuronal cells
Catalog No. 50-297-2328 Supplier Cayman Chemical Supplier No. 160604200 UG
A natural 14-naphthoquinone that has diverse effects in cells and animals causes the generation of ROS and induces apoptosis in cancer cells activates signaling through Nrf2 and ARE inducing the expression of Nrf2 target genes including NQO1 and HO-1 in cultured neuronal cells