Learn More
Aviva Systems Biology Corp PAF1 Antibody - N-terminal region (ARP61471_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61471_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PAF1. It was validated on Western Blot. . Alternate names: F23149_1, FLJ11123, PD2ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-388