Learn More
Aviva Systems Biology Corp PAFAH1B1 Antibody - N-terminal region (ARP56107_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56107_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PAFAH1B1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PAFAH1B1. Alternate names: LIS1, LIS2, MDCR, MDS, PAFAHApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61720