Learn More
Biomatik Corporation Pag 1 Polyclonal Antibody
Supplier:  Biomatik Corporation CAC09320
Encompass_Preferred
Pag 1 Polyclonal Antibody is a Bovine (Bos taurus) Polyclonal Antibody to PAG 1. The Pag 1 Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Bovine (Bos taurus). Immunogen: Recombinant Bovine Pregnancy-Associated Glycoprotein 1 Protein (54-380Aa). Applications: ELISA, WB. Target Synonyms: PAG 1. Accession Number: Q29432. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5279