Learn More
Grainger Supercool PAG Oil, 46 Viscosity, Synthetic, R134a, 6/Pk.
Catalog No. 502778092
Encompass_Preferred
This premium synthetic polyalkylene glycol (PAG) oil is formulated for R134a air conditioning systems. It features a 46 cSt viscosity and incorporates performance-enhancing additives and a U/V dye to assist in system maintenance.
  • Meets OEM performance specifications
  • Formulated with performance enhancers to improve heat transfer and increase system efficiency for reduced vent temperatures
  • Provides excellent lubricity to help reduce compressor wear
  • Contains conditioners, inhibitors, and anti-wear agents for enhanced compressor protection
  • Specially engineered for aftermarket air conditioning systems utilizing R134a refrigerant
Catalog No. 50-277-8092 Supplier Grainger Supplier No. 193Z62
This premium synthetic polyalkylene glycol (PAG) oil is formulated for R134a air conditioning systems. It features a 46 cSt viscosity and incorporates performance-enhancing additives and a U/V dye to assist in system maintenance.
  • Meets OEM performance specifications
  • Formulated with performance enhancers to improve heat transfer and increase system efficiency for reduced vent temperatures
  • Provides excellent lubricity to help reduce compressor wear
  • Contains conditioners, inhibitors, and anti-wear agents for enhanced compressor protection
  • Specially engineered for aftermarket air conditioning systems utilizing R134a refrigerant