Learn More
Genscript Corporation PAGE-MASTER Protein Standard (for SDS-PAGE)
Catalog No. NC1371685
Encompass
PAGE-MASTER Protein Standard (for SDS-PAGE) is a protein standard specially designed by GenScript. It consists of seven bands with molecular weight of 10 kD, 20 kD, 30 kD, 40 kD, 60 kD, 80 kD and 120 kD. All bands are highly purified, thus exhibit excellent performance in SDS-PAGE gels.
Catalog No. NC1371685 Supplier Genscript Corporation Supplier No. M00516
PAGE-MASTER Protein Standard (for SDS-PAGE) is a protein standard specially designed by GenScript. It consists of seven bands with molecular weight of 10 kD, 20 kD, 30 kD, 40 kD, 60 kD, 80 kD and 120 kD. All bands are highly purified, thus exhibit excellent performance in SDS-PAGE gels.