Learn More
Aviva Systems Biology Corp PAGE1 Antibody - middle region (ARP46092_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46092_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PAGE1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PAGE1. Alternate names: GAGE-9, GAGEB1, PAGE-1, AL5, CT16.3ApplicationWB. Species reactivity: Human
Catalog No. 50-014-59221