Learn More
Aviva Systems Biology Corp PAGE4 Antibody - middle region (ARP42336_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42336_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PAGE4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PAGE4. Alternate names: FLJ35184, GAGE-9, GAGEC1, JM27, PAGE-1, PAGE-4, JM-27, CT16.7ApplicationWB. Species reactivity: Human
Catalog No. 50-284-994