Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ PageRuler™ Prestained NIR Protein Ladder

Catalog No. p-7225571
Change view
Click to view available options
Quantity:
8 x 250 μL
2 x 250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI26635 2 x 250 mL
26-635-X4 8 x 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI26635 Supplier Thermo Scientific™ Supplier No. 26635
Only null left
Add to Cart
Edge
Add to Cart

A mixture of 10 proteins (11 to 250KDa) that are blue-stained and fluor-labeled for near-IR fluorescent visualization and protein sizing following electrophoresis.

The Thermo Scientific™ PageRuler Prestained NIR Protein Ladder is a mixture of 10 proteins (11 to 250KDa) that are blue-stained and fluor-labeled for near-IR fluorescent visualization and protein sizing following electrophoresis.

The protein MW markers in this ladder resolve into sharp bands when analyzed by SDS-PAGE and are labeled with a fluorescent dye for visualization with instruments equipped for detection of near-infrared (NIR) fluorescence. The bands are also directly visible because the proteins are prestained blue. The 55kDa band is of greater intensity and serves as a reference band. The protein ladder is conveniently packaged and ready to use: no heating, dilution or addition of a reducing agent is required before use.

Highlights:

  • Size range 10 proteins spanning 11 to 250kDa
  • Fluorescent detect using 670nm red laser or 700nm channels with fluorescence imagers
  • Ready-to-use supplied in a loading buffer for direct loading on gels
  • Sharp bands excellent sizing for near-infrared visualization of protein gels and blots
  • Reference band 55kDa band has greater intensity for easy orientation
  • Quality tested each lot evaluated by SDS-PAGE and Western blotting

Includes:

Proteins in 62.5mM Tris-H3PO4 (pH 7.5 at 25°C), 1mM EDTA, 2% SDS, 10mM DTT, 1mM NaN3 and 33% glycerol

Requires:

Fluorescent imager capable of near-infrared fluorescence detection, such as certain Typhoon™ Imagers and the LI-COR Odyssey™ Infrared Imaging System

Recommended for:

Protein sizing on SDS-polyacrylamide gels and Western blots for visualization with near-infrared (NIR) fluorescence imagers

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contents: two vials of 250 μL each

Storage buffer: 62.5 mM Tris-H3PO4 (pH 7.5 at 25°C), 1 mM EDTA, 2% SDS, 10 mM DTT, 1 mM NaN3 and 33% glycerol

Storage: Upon receipt store at -20°C

Color Blue
Detection Method Colorimetric, NIR Fluorescence (700 nm)
Number of Markers 10
Ready to Load Yes
Size Range 11 to 250 kDa
Gel Compatibility Bolt™ Bis-Tris Plus Gels, Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, SDS-PAGE Gels
Molecular Weight (g/mol) 250, 130, 95, 70, 55, 43, 34, 26, 15, 11 kDa
Quantity 2 x 250 mL
Shipping Condition Approved for shipment on Wet or Dry Ice
Product Line PageRuler
Product Type Protein Ladder
System Type Western Blotting, SDS-PAGE
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.