Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAH Mixture, 16 analytes various concentrations in acetonitrile/methanol (9:1), Ultra Scientific

Catalog No. USPM831A1
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-PM-831A-1 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-PM-831A-1 Supplier Agilent Technologies Supplier No. PM831A1
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Components:
  • acenaphthene 1000μg/mL
  • acenaphthylene 500μg/mL
  • anthracene 20μg/mL
  • benz[a]anthracene 50μg/mL
  • benzo[b]fluoranthene 20μg/mL
  • benzo[k]fluoranthene 20μg/mL
  • benzo[ghi]perylene 80μg/mL
  • benzo[a]pyrene 50μg/mL
  • chrysene 50μg/mL
  • dibenz[a,h]anthracene 200μg/mL
  • fluoranthene 50μg/mL
  • fluorene 100μg/mL
  • indeno[1,2,3-cd]pyrene 50μg/mL
  • naphthalene 500μg/mL
  • phenanthrene 40μg/mL
  • pyrene 100μg/mL
Matrix: 
  • acetonitrile/methanol (9:1)

Specifications

Chemical Name or Material PAH Mixture
CAS Max % 89.68, 9.96, 0.13, 0.06, 0.0
CAS 67-56-1, 83-32-9, 208-96-8, 120-12-7
Physical Form Liquid
Quantity 1 x 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.