Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAH Mixture (For EPA 8100/Appendix IX)

Catalog No. USPM8101
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-PM-8101 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-PM-8101 Supplier Agilent Technologies Supplier No. PM8101
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material PAH Mixture
Packaging Ampule
Methods For EPA Method 8100/Appendix IX
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) Each component in Methylene Chloride:
Acenaphthene    •1000μg/mL
Acenaphthylene    •1000μg/mL
Anthracene    •1000μg/mL
Benz[a]anthracene    •100μg/mL
Benzo[b]fluoranthene    •100μg/mL
Benzo[k]fluoranthene    •50μg/mL
Benzo[ghi]perylene    •100μg/mL
Benzo[a]pyrene    •100μg/mL
Chrysene    •100μg/mL
Dibenz[a,h]anthracene    •100μg/mL
Fluoranthene    •100μg/mL
Fluorene    •1000μg/mL
Indeno[1,2,3-cd]pyrene    •100μg/mL
Naphthalene    •1000μg/mL
Phenanthrene    •1000μg/mL
Pyrene    •100μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.