Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAH Mixture, Various concentrations in Dichloromethane, Ultra Scientific

Catalog No. USPM6121
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-PM-6121 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-PM-6121 Supplier Agilent Technologies Supplier No. PM6121
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Specifications

Chemical Name or Material PAH Mixture
CAS Max % 99.76, 75.0, 15.0, 0.75, 0.75, 0.15
CAS 83-32-9, 208-96-8, 120-12-7, 56-55-3, 205-99-2
Physical Form Liquid
Color Colorless
Quantity 1 x 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.