Learn More
Aviva Systems Biology Corp PAICS Antibody - N-terminal region (ARP46049_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46049_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PAICS. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PAICS. Alternate names: ADE2, ADE2H1, AIRC, DKFZp781N1372, MGC1343, MGC5024, PAISApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59197