Learn More
Research Products International Corp Pail/Drum Pump
SDP
Catalog No. 5000052446
Encompass_Preferred
Mess-Free Liquid Dispensing from Pails & Drums
Effortlessly dispense light oils and water-based fluids without spills or drips. Made from durable polypropylene, this dispenser features a flexible 33" intake tube and a 360° rotating spout for precise pouring. Compatible with Pail Lids with Spouts and drums with 3/4" or 2” openings. Supplied with a 2⅝" pail adapter and 3/4" & 2" bung adapters for versatile use. FDA compliant for safe handling.
Catalog No. 50-000-52446 Supplier Research Products International Corp Supplier No. 400201
Mess-Free Liquid Dispensing from Pails & Drums
Effortlessly dispense light oils and water-based fluids without spills or drips. Made from durable polypropylene, this dispenser features a flexible 33" intake tube and a 360° rotating spout for precise pouring. Compatible with Pail Lids with Spouts and drums with 3/4" or 2” openings. Supplied with a 2⅝" pail adapter and 3/4" & 2" bung adapters for versatile use. FDA compliant for safe handling.