Learn More
MSC Vikan Pail Lid, Polypropylene, Purple, For Bucket 5688
Catalog No. 502837551
Encompass
This pail lid is designed for Vikan Hygiene Bucket 5688, preventing foreign objects from falling in during storage. Its rim allows for secure stacking, optimizing space.
  • Prevents foreign objects from falling into buckets during storage.
  • Features a rim for secure bucket stacking.
  • Constructed from durable polypropylene in a purple color.
  • Complies with major food contact regulations (EC 1935/2004, EU 2023/2006/EC, FDA CFR 21).
  • Meets REACH Regulation (EC) No. 1907/2006, California Proposition 65, Halal, and Kosher standards.
  • Free from intentionally added PFAS.
  • Recyclable as polypropylene (PP - "5").
  • Operates across a wide temperature range, from -4 °F to 212 °F (usage) and sterilizable up to 249.8 °F.
  • Tolerates pH values from 2 to 10.5.
Catalog No. 50-283-7551 Supplier MSC Supplier No. 47172242
This pail lid is designed for Vikan Hygiene Bucket 5688, preventing foreign objects from falling in during storage. Its rim allows for secure stacking, optimizing space.
  • Prevents foreign objects from falling into buckets during storage.
  • Features a rim for secure bucket stacking.
  • Constructed from durable polypropylene in a purple color.
  • Complies with major food contact regulations (EC 1935/2004, EU 2023/2006/EC, FDA CFR 21).
  • Meets REACH Regulation (EC) No. 1907/2006, California Proposition 65, Halal, and Kosher standards.
  • Free from intentionally added PFAS.
  • Recyclable as polypropylene (PP - "5").
  • Operates across a wide temperature range, from -4 °F to 212 °F (usage) and sterilizable up to 249.8 °F.
  • Tolerates pH values from 2 to 10.5.