Learn More
MSC Vikan Lid for Bucket, Polypropylene, Purple, 12 Litre
Catalog No. 502837552
Encompass
This purple polypropylene lid is designed to prevent spillage from Hygiene Buckets and features a clip system for light security.
  • Prevents spillage and uses a clip system for light security.
  • Manufactured from durable polypropylene.
  • Purple color for easy identification.
  • Designed to fit 12 litre (3.17 gallon) buckets.
  • Complies with EU, FDA, and UK food contact material regulations, including (EC) 1935/2004 and FDA CFR 21.
  • Meets Halal and Kosher standards.
  • Free from intentionally added PFAS.
  • Recyclable as polypropylene (PP).
Catalog No. 50-283-7552 Supplier MSC Supplier No. 47172333
This purple polypropylene lid is designed to prevent spillage from Hygiene Buckets and features a clip system for light security.
  • Prevents spillage and uses a clip system for light security.
  • Manufactured from durable polypropylene.
  • Purple color for easy identification.
  • Designed to fit 12 litre (3.17 gallon) buckets.
  • Complies with EU, FDA, and UK food contact material regulations, including (EC) 1935/2004 and FDA CFR 21.
  • Meets Halal and Kosher standards.
  • Free from intentionally added PFAS.
  • Recyclable as polypropylene (PP).