Learn More
MSC Vikan Lid for Bucket, Polypropylene, White, Food Contact Compliant
Catalog No. 502837636
Encompass
This white polypropylene lid is designed for hygiene buckets, offering secure closure to prevent spillage and inhibit bacterial growth. Ideal for food manufacturing, retail, and food service, it complies with stringent food contact material regulations, ensuring safe use in critical hygienic environments.
  • Purpose-built for critical hygiene and food safety applications.
  • Rapid draining design helps prevent bacteria growth.
  • Snap-on for secure storage and spillage prevention.
  • Stackable and lightweight for efficient handling.
  • Resists harsh chemicals and cleaning agents.
  • Easy to clean and maintain.
  • Durable for long-lasting daily use.
  • Includes a hanging hole for convenient storage.
  • Complies with food contact material regulations (EC) 1935/2004 and FDA 21 CFR.
Catalog No. 50-283-7636 Supplier MSC Supplier No. 52583531
This white polypropylene lid is designed for hygiene buckets, offering secure closure to prevent spillage and inhibit bacterial growth. Ideal for food manufacturing, retail, and food service, it complies with stringent food contact material regulations, ensuring safe use in critical hygienic environments.
  • Purpose-built for critical hygiene and food safety applications.
  • Rapid draining design helps prevent bacteria growth.
  • Snap-on for secure storage and spillage prevention.
  • Stackable and lightweight for efficient handling.
  • Resists harsh chemicals and cleaning agents.
  • Easy to clean and maintain.
  • Durable for long-lasting daily use.
  • Includes a hanging hole for convenient storage.
  • Complies with food contact material regulations (EC) 1935/2004 and FDA 21 CFR.