This green polypropylene lid is specifically designed for Vikan Hygiene Bucket 5688, ensuring contents are protected from foreign objects. Its innovative design allows buckets to be stacked efficiently, maximizing storage. Ideal for laboratory and industrial use, this lid meets stringent food safety and regulatory compliance standards.
- Prevents foreign objects from falling into the bucket when used for storage.
- Rim in the middle allows buckets to be stacked securely.
- Made from durable polypropylene.
- Complies with EC 1935/2004 for food contact materials.
- Complies with FDA Regulation CFR 21.
- Halal and Kosher compliant.
- Suitable for usage in temperatures ranging from -20 °C to 100 °C.
- Can be cleaned in a dishwasher up to 93 °C.
- Sterilizable via autoclave at 121 °C.