Learn More
MSC Vikan Lid for Bucket, Polypropylene, Red, Stackable, FDA Compliant
Catalog No. 502836258
Encompass
This lid is designed for Vikan Hygiene Bucket 5688x, preventing foreign objects from entering the bucket during storage. Its unique rim design also enables secure stacking of buckets.
  • Prevents foreign object contamination
  • Allows for stackable storage
  • Constructed from polypropylene
  • Red color for easy identification
  • Complies with food contact material regulations (EC) 1935/2004
  • Manufactured according to EU Regulation 2023/2006/EC of Good Manufacturing Practice
  • Complies with FDA Regulation CFR 21
  • Complies with UK 2019 No. 704 on food contact materials
  • Meets REACH Regulation (EC) No. 1907/2006
  • Complies with California Proposition 65
  • Halal and Kosher compliant
  • No intentionally added PFAS
  • Recyclable as polypropylene
Catalog No. 50-283-6258 Supplier MSC Supplier No. 47172283
This lid is designed for Vikan Hygiene Bucket 5688x, preventing foreign objects from entering the bucket during storage. Its unique rim design also enables secure stacking of buckets.
  • Prevents foreign object contamination
  • Allows for stackable storage
  • Constructed from polypropylene
  • Red color for easy identification
  • Complies with food contact material regulations (EC) 1935/2004
  • Manufactured according to EU Regulation 2023/2006/EC of Good Manufacturing Practice
  • Complies with FDA Regulation CFR 21
  • Complies with UK 2019 No. 704 on food contact materials
  • Meets REACH Regulation (EC) No. 1907/2006
  • Complies with California Proposition 65
  • Halal and Kosher compliant
  • No intentionally added PFAS
  • Recyclable as polypropylene