Learn More
MSC Vikan Lid, Polypropylene, 6 Litre, Lime
Catalog No. 502836730
Encompass
This polypropylene lid is designed for the Vikan Hygiene Bucket 5688, preventing foreign objects from falling into the bucket during storage. Its rim allows for efficient stacking of buckets.
  • Purpose-built for food manufacturing, food retail, restaurants, and food service where hygiene and food safety are critical.
  • Snap-on lid provides secure storage.
  • Features a unique hanging hole for easy storage.
  • Stackable design for efficient storage.
  • Designed to drain rapidly to help prevent bacteria growth.
  • Withstands harsh chemicals and cleaning agents.
  • Colour-coded for use with hygienic zoning plans and 5S lean programmes.
  • Easy to clean and maintain for hygiene control.
  • Durable construction provides long-lasting performance with daily use.
  • Lightweight design reduces user fatigue.
Catalog No. 50-283-6730 Supplier MSC Supplier No. 47172259
This polypropylene lid is designed for the Vikan Hygiene Bucket 5688, preventing foreign objects from falling into the bucket during storage. Its rim allows for efficient stacking of buckets.
  • Purpose-built for food manufacturing, food retail, restaurants, and food service where hygiene and food safety are critical.
  • Snap-on lid provides secure storage.
  • Features a unique hanging hole for easy storage.
  • Stackable design for efficient storage.
  • Designed to drain rapidly to help prevent bacteria growth.
  • Withstands harsh chemicals and cleaning agents.
  • Colour-coded for use with hygienic zoning plans and 5S lean programmes.
  • Easy to clean and maintain for hygiene control.
  • Durable construction provides long-lasting performance with daily use.
  • Lightweight design reduces user fatigue.