Learn More
Grainger Vikan Bucket Lid, Polypropylene, 12 in. Diameter, Orange, 1 ea.
Catalog No. 502786905
Encompass_Preferred
This snap-on polypropylene lid is designed to secure 12 L (3 gal.) hygiene buckets, providing protection against contaminants while preventing spills during transport. Its durable, food-grade construction is suitable for rigorous environments including food manufacturing, food service, and retail. The lid features a color-coded design to support hygienic zoning plans and 5S lean programs, and is engineered to facilitate rapid drainage to minimize microbial growth.
  • Snap-on design ensures secure closure and leakproof transport
  • Color-coded to support hygienic zoning and facility organization
  • Constructed from durable, food-grade polypropylene
  • Withstands harsh chemicals and cleaning agents
  • Autoclavable up to 121°C for thorough sterilization
  • Compliant with FDA and EU food contact regulations
Catalog No. 50-278-6905 Supplier Grainger Supplier No. 18G943
This snap-on polypropylene lid is designed to secure 12 L (3 gal.) hygiene buckets, providing protection against contaminants while preventing spills during transport. Its durable, food-grade construction is suitable for rigorous environments including food manufacturing, food service, and retail. The lid features a color-coded design to support hygienic zoning plans and 5S lean programs, and is engineered to facilitate rapid drainage to minimize microbial growth.
  • Snap-on design ensures secure closure and leakproof transport
  • Color-coded to support hygienic zoning and facility organization
  • Constructed from durable, food-grade polypropylene
  • Withstands harsh chemicals and cleaning agents
  • Autoclavable up to 121°C for thorough sterilization
  • Compliant with FDA and EU food contact regulations