Learn More
MSC Vestil Pail Liner, Polyethylene, 5 Gallon, FDA Compliant
Catalog No. 502835838
Encompass
Pail liners are designed to protect and extend the life of plastic and steel 5-gallon pails. They create a barrier, preventing damage to the pails and simplifying material disposal. These liners are constructed for use in chemical, cosmetic, food & beverage, and pharmaceutical applications, contributing to a cleaner environment by maximizing product recovery.
  • Unique design protects and extends the life of plastic and steel 5-gallon pails.
  • Prevents damage to pails by creating a barrier.
  • Constructed to handle chemical, cosmetic, food & beverage and pharmaceutical applications.
  • FDA compliant semi-rigid polyethylene resins.
Catalog No. 50-283-5838 Supplier MSC Supplier No. 81734030
Pail liners are designed to protect and extend the life of plastic and steel 5-gallon pails. They create a barrier, preventing damage to the pails and simplifying material disposal. These liners are constructed for use in chemical, cosmetic, food & beverage, and pharmaceutical applications, contributing to a cleaner environment by maximizing product recovery.
  • Unique design protects and extends the life of plastic and steel 5-gallon pails.
  • Prevents damage to pails by creating a barrier.
  • Constructed to handle chemical, cosmetic, food & beverage and pharmaceutical applications.
  • FDA compliant semi-rigid polyethylene resins.