Learn More
MSC Polar Tech Hazardous Shipper, 2x1 gal., 1 Kit
Catalog No. 501915881
Encompass
This Hazardous Materials FStyle Can Foam Shipper Kit is designed for safely shipping solvents, thinners, and paints. It is approved for UN/USA D.O.T. standards and accepted by both domestic and international carriers, providing a reliable solution for hazardous material transport.
  • Holds two 1 gal. F-style cans
  • Includes a pre-assembled carton, F-style can with cap, and foam inserts
  • Constructed from corrugated, unlined metal, and HDPE materials
Catalog No. 50-191-5881 Supplier MSC Supplier No. 39554050
This Hazardous Materials FStyle Can Foam Shipper Kit is designed for safely shipping solvents, thinners, and paints. It is approved for UN/USA D.O.T. standards and accepted by both domestic and international carriers, providing a reliable solution for hazardous material transport.
  • Holds two 1 gal. F-style cans
  • Includes a pre-assembled carton, F-style can with cap, and foam inserts
  • Constructed from corrugated, unlined metal, and HDPE materials