Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Paint Masking Tape

Catalog No. 19070908 Shop All 3M Products
Change view
Click to view available options
Material:
Crepe Paper
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-070-908 Crepe Paper
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-070-908 Supplier 3M™ Supplier No. 7000088516
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3M Company Paint Masking Tape provides strong tack and adhesion and resists lifting

  • Clean removal from many surfaces following multi-bake or high temperature bake
  • Smooth mini-crepe paper backing conforms and maintains integrity when formed around a corner
  • Strong backing is easy to tear, yet resists slivering and provides one-piece removal
  • Controlled unwind dispenses evenly, reducing fatigue
  • High-performance crepe paper masking tape designed to meet aerospace needs in most painting applications, including the use of multiple bake cycles
  • 48 mm x 55 m 7.6 mil
  • 3M Catalog# 231

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Protect from paint overspray; Masking body jig barcode; On-line paint repair masking
Color Tan
Material Crepe Paper
Model 231
Quantity 24 Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.