Learn More
Aviva Systems Biology Corp PAIP1 Antibody - C-terminal region (ARP40651_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40651_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PAIP1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human PAIP1. Alternate names: ApplicationIHC, WB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57573