Learn More
Aviva Systems Biology Corp PAIP2 Antibody - N-terminal region (ARP56241_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56241_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PAIP2. It was validated on Western Blot. . Alternate names: MGC72018, PAIP2AApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-128