Learn More
Fritsch Milling And Sizing Inc Pair Of Lateral Support Walls Made Of Zirconium Oxide, P-1 Cl Size 1
SDP
Catalog No. 501534381
Encompass
Lateral support walls made of zirconium oxide for the FRITSCH Jaw Crusher PULVERISETTE 1 classic line, Size 1.
General material specification:
Material: zirconium Oxide
Abrasion resistance: good
Suitable for: hard, brittle samples
Catalog No. 50-153-4381 Supplier Fritsch Milling And Sizing Inc Supplier No. 43.0160.27
Add to Cart
Add to Cart
Request a Quote
Lateral support walls made of zirconium oxide for the FRITSCH Jaw Crusher PULVERISETTE 1 classic line, Size 1.
General material specification:
Material: zirconium Oxide
Abrasion resistance: good
Suitable for: hard, brittle samples