Learn More
Carnabio Usa Inc PAK1/5UG/FULL-LENGTH
Supplier:  Carnabio Usa Inc 071235
Encompass
PAK1, Full-length, Wild type, Amino Acid 1-545, NP_002567.3, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0553