Learn More
Eurofins Discoverx PAK1, active, 250 µg. (14-927M)
Supplier:  Eurofins Discoverx 14927M
Encompass
N-terminal FLAG-tagged recombinant, human PAK1 amino acids 150-end. This protein is a family member of serine/threonine p21-activating kinases, known as PAK proteins. These proteins are critical effectors that link RhoGTPases to cytoskeleton reorganization and nuclear signaling, and they serve as targets for the small GTP binding proteins Cdc42 and Rac. This specific family member regulates cell motility and morphology. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9428