Learn More
Eurofins Discoverx PAK2, active, 10 µg. (14-481)
Supplier:  Eurofins Discoverx 14481
Encompass
N-terminal 6xHis and T7, and C-terminal 6xHis-tagged recombinant, human PAK2, amino acids 3-end. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8853