Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human PAK2 (PAK65), GST Tag Recombinant Protein

Catalog No. PV4565
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV4565 10 μg
PV4566 1 mg
PR8265A 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PV4565 Supplier Invitrogen™ Supplier No. PV4565
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length.

PAK2 (serine/threonine-protein kinase PAK2) is a serine/threonine kinase that plays a role in a variety of different signaling pathways including cytoskeleton regulation, cell motility, cell cycle progression, apoptosis, or proliferation. It acts as a downstream effector of the small GTPases CDC42 and RAC1. Activation by the binding of active CDC42 and RAC1 results in a conformation change and a subsequent autophosphorylation on several serine and/or threonine residues. Full-length PAK2 stimulates cell survival and cell growth. PAK proteins are critical effectors that link Rho GTPases to cytoskeleton reorganization and nuclear signaling. PAK proteins, a family of serine/threonine p21-activating kinases, serve as targets for the small GTP binding proteins Cdc42 and RAC and have been implicated in a wide range of biological activities.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q13177
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris with 0.02% Triton X-100, 50% glycerol, 0.5mM EDTA, 2mM DTT, 150mM NaCl and no preservative; pH 7.5
Gene ID (Entrez) 5062
Molecular Weight (g/mol) 85.5 kDa
Name Human PAK2 (PAK65), GST Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 5330420P17Rik; A130002K10Rik; AI836325; Alpha-PAK; Beta-PAK; C-t-PAK2; D16Ertd269e; gamma pak; gamma-PAK; mKIAA4182; Oligophrenin-3; OPHN3; p21 (CDKN1A)-activated kinase 2; p21 (RAC1) activated kinase 2; p21 protein (Cdc42/Rac)-activated kinase 2; p21-activated kinase 2; p27; p34; p58; p65-PAK; PAK alpha; PAK beta; PAK gamma; PAK1; PAK-1; Pak2; PAK-2; PAK-2p27; PAK-2p34; PAK3; PAK-3; PAK65; PAKgamma; S6/H4 kinase; serine/threonine-protein kinase PAK 2
Common Name PAK2
Gene Symbol Pak2
Kinase Family STE20 Kinase Family
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Assay Entry Biochemical phospho-specific activity (no antibody)
Kinase Group Ser⁄Thr Kinases: STE Group
Shipping Condition Dry Ice
System Compatibility Z'-LYTE™, Omnia™ Bulk Substrate
Target Entry PAK2
Target Gene protein kinase N2
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Full Length
Form Liquid
Protein Subtype Serine/Threonine Kinases
Research Category Signal Transduction
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.