Learn More
Sino Biological Recombinant Human TrkC Protein (L686M, His Tag), Active
Supplier: Sino Biological N1812DH500
Recombinant human TRKC (L686M) (507-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.