Learn More
Eurofins Discoverx PAK3, active, 10 µg. (14-683)
Supplier:  Eurofins Discoverx 14683
Encompass
C-terminal 6xHis-tagged recombinant, human full length PAK3. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9181