Learn More
Eurofins Discoverx PAK4, active, 250 µg. (14-584M)
Supplier:  Eurofins Discoverx 14584M
Encompass
N-terminal 6xHis and N-terminal GST-tagged recombinant, human PAK4 amino acids 295-end. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9008