Learn More
Eurofins Discoverx PAK5, active, 250 µg. (14-699M)
Supplier:  Eurofins Discoverx 14699M
Encompass
N-terminal 6xHis-tagged recombinant, human PAK5 amino acids 425-end. PAK5 is also known as PAK7. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9210