Learn More
Eurofins Discoverx PAK6, active, 10 µg. (14-633)
Supplier:  Eurofins Discoverx 14633
Encompass
N-terminal 6xHis-tagged recombinant, human PAK6 amino acids 382-end. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9091