Learn More
Trajan Scientific Pal 10Ml Clear Glass Vial 18Mm
Catalog No. 501920901
Encompass
PAL System Vial 10CV 10ml Clear Glass with Label designed for the PAL Autosampler. 46x22.5mm 1st Class Hydrolytic Glass with flat finish for better sealing fits ND18 Screw Caps Pk of 100 Pcs
Catalog No. 501920901 Supplier Trajan Scientific Supplier No. VIAL10ND18CG100
PAL System Vial 10CV 10ml Clear Glass with Label designed for the PAL Autosampler. 46x22.5mm 1st Class Hydrolytic Glass with flat finish for better sealing fits ND18 Screw Caps Pk of 100 Pcs