Learn More
Trajan Scientific Pal 20Ml Amber Glass Vial 18Mm
Catalog No. 501920902
Encompass
PAL System Vial 20CV 20ml Amber Glass with Label designed for the PAL Autosampler. 75.5x22.5mm 1st Class Hydrolytic Glass with flat finish for better sealing fits ND18 Screw Caps Pk of 100 Pcs
Catalog No. 501920902 Supplier Trajan Scientific Supplier No. VIAL20ND18AG100
PAL System Vial 20CV 20ml Amber Glass with Label designed for the PAL Autosampler. 75.5x22.5mm 1st Class Hydrolytic Glass with flat finish for better sealing fits ND18 Screw Caps Pk of 100 Pcs