Learn More
Trajan Scientific Pal 20Ml Clear Glass Vial 18Mm
Catalog No. 501920903
Encompass
PAL System Vial 20CV 20ml Clear Glass with Label designed for the PAL Autosampler. 75.5x22.5mm 1st Class Hydrolytic Glass with flat finish for better sealing fits ND18 Screw Caps Pk of 100 Pcs
Catalog No. 501920903 Supplier Trajan Scientific Supplier No. VIAL20ND18CG100
PAL System Vial 20CV 20ml Clear Glass with Label designed for the PAL Autosampler. 75.5x22.5mm 1st Class Hydrolytic Glass with flat finish for better sealing fits ND18 Screw Caps Pk of 100 Pcs