Learn More
Trajan Scientific Pal 2Ml Amber Glass Vial 9Mm
Catalog No. 501920896
Encompass
PAL System Vial 2CV 1.5ml Amber Glass with Label designed for the PAL Autosampler. 12x32mm 1st Class Hydrolytic Glass fits ND9 Screw Caps Pk of 100 Pcs
Catalog No. 501920896 Supplier Trajan Scientific Supplier No. VIAL1.5ND9AG100
PAL System Vial 2CV 1.5ml Amber Glass with Label designed for the PAL Autosampler. 12x32mm 1st Class Hydrolytic Glass fits ND9 Screw Caps Pk of 100 Pcs