Learn More
Trajan Scientific Pal 2Ml Pp Vial 9Mm
Catalog No. 501920898
Encompass
PAL System Vial 2CV 1.5mL Clear PP designed for the PAL Autosampler. 12x32mm with filling lines fits ND9 Screw Caps Pk of 100 Pcs
Catalog No. 501920898 Supplier Trajan Scientific Supplier No. VIAL1.5ND9PP100
PAL System Vial 2CV 1.5mL Clear PP designed for the PAL Autosampler. 12x32mm with filling lines fits ND9 Screw Caps Pk of 100 Pcs