Learn More
Bachem Palmitoyl-Gly-His-Lys-OH
SDP
Catalog No. NC2404497
Encompass_Preferred
Pal-GHK or Palmitoyl Tripeptide-1 is a version of GHK peptide (copper-binding peptide) designed for better skin penetration, since it is a messenger peptide for collagen renewal. Therefore Pal-GHK leads to wrinkle reduction, skin firming and skin improvement.
Catalog No. NC2404497 Supplier Bachem Supplier No. 4142828.2004
Pal-GHK or Palmitoyl Tripeptide-1 is a version of GHK peptide (copper-binding peptide) designed for better skin penetration, since it is a messenger peptide for collagen renewal. Therefore Pal-GHK leads to wrinkle reduction, skin firming and skin improvement.