Learn More
Medchemexpress LLC Pal-Glu OSu -OtBu 10g
SDP
Catalog No. 5000206862
Encompass_Preferred
Pal-Glu(OSu)-OtBu (Palmitoyl-Glu(OSu)-OBut) is an important intermediate in the synthesis process of Liraglutide (HY-P0014)[1]
Catalog No. 50-002-06862 Supplier Medchemexpress LLC Supplier No. HYW09957810G
Pal-Glu(OSu)-OtBu (Palmitoyl-Glu(OSu)-OBut) is an important intermediate in the synthesis process of Liraglutide (HY-P0014)[1]