Learn More
Cayman Chemical Pal-KTTKS (acetate), Purity: ≥90%, 100 mg
SDP
Catalog No. 502937444
Encompass_Preferred
A lipidated pentapeptide consisting of a fragment of the type I collagen C-terminal propeptide conjugated to palmitic acid; increases collagen production in human corneal and dermal fibroblasts at 0.002, 0.004, and 0.008 wt%; found in the stratum corneum, epidermis, and dermis of isolated hairless mouse skin following topical administration at 50 μg/cm2; can self-assemble into flat tapes and extended fibrillar structures; has been detected in anti-wrinkle creams
Catalog No. 50-293-7444 Supplier Cayman Chemical Supplier No. 27060100 MG
A lipidated pentapeptide consisting of a fragment of the type I collagen C-terminal propeptide conjugated to palmitic acid; increases collagen production in human corneal and dermal fibroblasts at 0.002, 0.004, and 0.008 wt%; found in the stratum corneum, epidermis, and dermis of isolated hairless mouse skin following topical administration at 50 μg/cm2; can self-assemble into flat tapes and extended fibrillar structures; has been detected in anti-wrinkle creams